Learn More
R&D Systems™ Proteome Purify 2 Human Serum Protein Immunodepletion Resin
A single-use resin that depletes serum or plasma of albumin and IgG to improve visualization of target proteins
Brand: R&D Systems™ IDR002-040
Additional Details : Weight : 0.58200kg
Includes: Immunodepletion Resin, Spin-X Filter Units
Description
Allows user to increase protein load or concentration
Removal allows visualization of proteins that co-migrate or co-purify with the high-abundance proteins
Kit removes proteins representing approximately 60% of the protein contained in serum and plasma
Specifications
40 Tests | |
Immunodepletion Resin | |
Store the unopened product at 2 - 8° C. Do not use past expiration date. |