missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Proteome Purify 2 Human Serum Protein Immunodepletion Resin

A single-use resin that depletes serum or plasma of albumin and IgG to improve visualization of target proteins

Brand:  R&D Systems™ IDR002-020

Additional Details : Weight : 0.58200kg

 View more versions of this product

Product Code. 18482070

  • £517.27 / 20mL
Estimated Shipment: 05-08-2024
to see stock.

Includes: Immunodepletion Resin, Spin-X Filter Units

Description

Description

Allows user to increase protein load or concentration
Removal allows visualization of proteins that co-migrate or co-purify with the high-abundance proteins
Kit removes proteins representing approximately 60% of the protein contained in serum and plasma

Specifications

Specifications

20 Tests
Immunodepletion Resin
Store the unopened product at 2 - 8° C. Do not use past expiration date.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers