missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ Protino™ Ni-IDA Resin
For Purification of Polyhistidine-Tagged Proteins, Batch Binding, Gravity Flow Column Chromatography, MPLC/FPLC Applications
£46.60 - £468.00
Specifications
Binding Capacity | 20mg/g |
---|---|
Binding Property | Batch |
Kit Contents | Buffers |
Matrix | Macroporous Silica |
Max. Operating Pressure | 145 psi |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11932532
|
Macherey-Nagel™
745210.5 |
5 g Dry Silica Bulk Resin |
£46.60
5g |
In Stock
Log in to see stock. |
|||||
11972412
|
Macherey-Nagel™
745210.30 |
30 g Dry Silica Bulk Resin |
£468.00
30g |
In Stock
Log in to see stock. |
|||||
Description
Protino™ Ni-IDA, the purer alternative for higher yields of His-tag proteins- Chelating group IDA, three protein binding sites
- High protein yield/recovery even from diluted samples
- High protein concentration
- High binding specificity, less unspecific binding of contaminating proteins compared to other common IMAC matrices
- Dry resin, storage at room temperature
- Ready to use pre-packed columns filled with dry Protino™ Ni-IDA resin
- Buffers included
- Dry bulk matrix
- Rigid support silica matrix, compatible with high pressure and high flow rates, does not shrink or swell - highest flexibility
- Batch procedure, gravity flow column chromatography, FPLC™ applications
- Empty Protino™ columns available for use with Protino™ Ni-IDA resin
- Empty 96 well receiver plates available for use with Protino™ Ni-IDA resin
- Ready to use pre-packed 96 well plates filled with dry Protino™ Ni-IDA resin
- Buffers included For purification of polyhistidine-tagged proteins, batch binding, gravity flow column chromatography, MPLC/FPLC™ applications.
Specifications
20mg/g | |
Buffers | |
145 psi | |
Resin |
Batch | |
Macroporous Silica | |
For Purification of Polyhistidine-Tagged Proteins, Batch Binding, Gravity Flow Column Chromatography, MPLC/FPLC Applications | |
IDA (iminodiacetic acid) |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Danger
Hazard Category
- Skin sensitiser Category 1
- Respiratory sensitiser Category 1
- Germ cell mutagenicity Category 2
- Carcinogenicity Category 1A
- LONG-TERM AQUATIC HAZARD Chronic 1
Hazard Statement
- H317-May cause an allergic skin reaction.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H341-Suspected of causing genetic defects.
- H350i-May cause cancer by inhalation.
- H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
- P201-Obtain special instructions before use.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- P391-Collect spillage.
- P405-Store locked up.
Supplemental information
- MIXTURE LIST-Contains : resin with Ni in nickel(II) complexes