missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Protino Ni-TED Resin for His-tag Protein Purification

Product Code. 11922532
Click to view available options
Quantity:
1 Each
120 g Dry Silica Bulk Resin
30 g Dry Silica Bulk Resin
5 g Dry Silica Bulk Resin
Unit Size:
120g
30g
5g
600g
This item is not returnable. View return policy

Product Code. 11922532

Brand: Macherey-Nagel™ 745200.5

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For Purification of Polyhistidine-Tagged Proteins, Batch Binding, Gravity Flow Column Chromatography, MPLC/FPLC Applications. The matrix of choice for highest protein purity as dry bulk material.

Protino™ NI-TED, the purer alternative for His-tag protein purification
  • Chelating group TED, one protein binding site
  • High stability against reducing/chelating agents
  • Low metal leaching
  • High protein purity
  • High binding specificity, less unspecific binding of contaminating proteins compared to other common IMAC matrices
  • Dry resin, storage at room temperature
Protino™ NI-TED packed columns
  • Ready to use pre-packed columns filled with dry Protino™ Ni-TED resin
  • Buffers included
Protino™ NI-TED resin
  • Dry bulk matrix
  • Rigid support silica matrix, compatible with high pressure and high flow rates, does not shrink well - highest flexibility
  • Batch procedure, gravity flow column chromatography, FPLC™ applications
  • Empty Protino™ columns available for use with Protino™ Ni-TED resin
  • Empty 96 well receiver plates available for use with Protino™ Ni-TED resin For purification of polyhistidine-tagged proteins; batch binding, gravity flow column chromatography, MPLC/FPLC™ applications.

Specifications

Binding Capacity 10mg/g
Binding Property Batch
Flow Rate 0.5 to 1.0 mL/min./cm2
Kit Contents Buffers
Matrix Macroporous Silica
Max. Operating Pressure 145 psi
Quantity 5 g Dry Silica Bulk Resin
For Use With (Application) For Purification of Polyhistidine-Tagged Proteins, Batch Binding, Gravity Flow Column Chromatography, MPLC/FPLC Applications
Type Resin
Ligand Type TED (tris-carboxymethyl ethylene diamine)
compliance-icons
Hazard Category
  • Respiratory sensitiser Category 1
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.