Learn More
Tocris Bioscience™ PRX 07034
Potent and selective 5-HT6 receptor antagonist (IC50 = 19 nM, Ki = 4 to 8 nM)
Brand: Tocris Bioscience™ 6748/10
This item is not returnable.
View return policy
Description
PRX 07034 is a potent and selective 5-HT6 receptor antagonist (IC50 = 19 nM, Ki = 4 - 8 nM). Enhances working memory in rats.Specifications
N-(1-(5-Chloro-2,3-dimethoxyphenyl)ethyl)-2-(methylsulfonyl)-5-(piperazin-1-yl)aniline hydrochloride | |
10 mg | |
490.44 |
903580-39-2 | |
C21H28ClN3O4S.HCl | |
98% |