missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ PRX 07034
Potent and selective 5-HT6 receptor antagonist (IC50 = 19 nM, Ki = 4 to 8 nM)
£163.06 - £638.21
Specifications
Chemical Name or Material | N-(1-(5-Chloro-2,3-dimethoxyphenyl)ethyl)-2-(methylsulfonyl)-5-(piperazin-1-yl)aniline hydrochloride |
---|---|
CAS | 903580-39-2 |
Molecular Formula | C21H28ClN3O4S.HCl |
Formula Weight | 490.44 |
Purity | 98% |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18715406
|
Tocris Bioscience™
6748/10 |
10 mg |
£163.06
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18739226
|
Tocris Bioscience™
6748/50 |
50 mg |
£638.21
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Enhances working memory in rats.Specifications
N-(1-(5-Chloro-2,3-dimethoxyphenyl)ethyl)-2-(methylsulfonyl)-5-(piperazin-1-yl)aniline hydrochloride | |
C21H28ClN3O4S.HCl | |
98% |
903580-39-2 | |
490.44 |
Safety and Handling
Recommended Storage : Store at -20°C