Learn More
PSI-7409 tetrasodium, MedChemExpress
MedChemExpress PSI-7409 tetrasodium is an active 5'-triphosphate metabolite of sofosbuvir (PSI-7977), inhibiting HCV NS5B polymerases, with IC50s of 1.6, 2.8, 0.7 and 2.6 μM for GT 1b_Con1, GT 2a_JFH1, GT 3a, and GT 4a NS5B polymerases, respectively.
Brand: Medchem Express HY-15745A
Description
PSI-7409 tetrasodium is an active 5'-triphosphate metabolite of sofosbuvir (PSI-7977), inhibiting HCV NS5B polymerases, with IC50s of 1.6, 2.8, 0.7 and 2.6 μM for GT 1b_Con1, GT 2a_JFH1, GT 3a, and GT 4a NS5B polymerases, respectively.Specifications
PSI-7409 tetrasodium | |
White | |
C10H12FN2Na4O14P3 | |
H2O : ≥ 125 mg/mL (212.55 mM) | |
588.09 | |
95.0% | |
Solid |
1621884-22-7 | |
Research | |
25 mg | |
O[C@@H]([C@@](C)(F)[C@H](N1C(NC(C=C1)=O)=O)O2)[C@H]2COP(OP(OP(O[Na])(O[Na])=O)(O[Na])=O)(O[Na])=O | |
588.09 | |
Research | |
COVID-19-anti-virus |