Learn More
PSI-7409 tetrasodium, MedChemExpress
MedChemExpress PSI-7409 tetrasodium is an active 5'-triphosphate metabolite of sofosbuvir (PSI-7977), inhibiting HCV NS5B polymerases, with IC50s of 1.6, 2.8, 0.7 and 2.6 μM for GT 1b_Con1, GT 2a_JFH1, GT 3a, and GT 4a NS5B polymerases, respectively.

£312.18 - £2883.43
Specifications
Chemical Name or Material | PSI-7409 tetrasodium |
---|---|
CAS | 1621884-22-7 |
Color | White |
Purity Grade Notes | Research |
Molecular Formula | C10H12FN2Na4O14P3 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16496788
![]() |
Medchem Express
HY-15745A |
10 mg |
£1442.32
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16493486
![]() |
Medchem Express
HY-15745A |
25 mg |
£2883.43
25mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16479337
![]() |
Medchem Express
HY-15745A |
5 mg |
£937.75
5mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16461549
![]() |
Medchem Express
HY-15745A |
1 mg |
£312.18
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PSI-7409 tetrasodium is an active 5'-triphosphate metabolite of sofosbuvir (PSI-7977), inhibiting HCV NS5B polymerases, with IC50s of 1.6, 2.8, 0.7 and 2.6 μM for GT 1b_Con1, GT 2a_JFH1, GT 3a, and GT 4a NS5B polymerases, respectively.Specifications
PSI-7409 tetrasodium | |
White | |
C10H12FN2Na4O14P3 | |
O[C@@H]([C@@](C)(F)[C@H](N1C(NC(C=C1)=O)=O)O2)[C@H]2COP(OP(OP(O[Na])(O[Na])=O)(O[Na])=O)(O[Na])=O | |
588.09 | |
Research | |
COVID-19-anti-virus |
1621884-22-7 | |
Research | |
H2O : ≥ 125 mg/mL (212.55 mM) | |
588.09 | |
95.0% | |
Solid |
Safety and Handling
missing translation for 'shelfLife' : -20°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Recommended Storage : -20°C, sealed storage, away from moisture∣In solvent : -80°C, 6 months∣-20°C, 1 month (sealed storage, away from moisture)
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.