missing translation for 'onlineSavingsMsg'
Learn More

Kartell™ PTFE Beaker

Possess excellent chemical, mechanical and heat resistance. Kartell™ PTFE Beaker is a non-contaminating beaker that is completely inert to highly corrosive reagents.

£31.25 - £195.00

Specifications

Color White
Material PTFE
For Use With (Application) Suitable for Foodstuff
Temperature (Metric) Max. Operating 250°C
Type Beaker
View More Specs

Products 5
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
10624242
View Documents Promotion Details
Kartell™
0071700
50 mL
£31.25
Each
Estimated Shipment: 29-11-2024
to see stock.
Add to basket
 
10081081
View Documents Promotion Details
Kartell™
0071800
100 mL
£40.00
Each
In Stock
to see stock.
Edge
Add to basket
 
10366111
View Documents Promotion Details
Kartell™
0072000
250 mL
£64.10
Each
Estimated Shipment: 29-11-2024
to see stock.
Add to basket
 
10122811
View Documents Promotion Details
Kartell™
0072100
500 mL
£87.35
Each
Estimated Shipment: 23-01-2025
to see stock.
Add to basket
 
10081091
View Documents Promotion Details
Kartell™
0072300
1000 mL
£195.00
Each
Estimated Shipment: 23-01-2025
to see stock.
Add to basket
 
Description

Description

  • Excellent chemical, mechanical, and heat resistance
  • Will withstand temperatures continuously up to 250°C
  • Will withstand temperatures for short intervals up to 300°C
  • Suitable for applications with food
Specifications

Specifications

White
Suitable for Foodstuff
Beaker
PTFE
250°C
No
Videos
SDS
Documents

Documents

Product Certifications