Learn More
Bohlender™ PTFE Blunt Tip Tweezers
Use these tweezers for a variety of applications. BOLA PTFE Tweezers feature universal chemical resistance.
Brand:  Bohlender™ H912-06
 View more versions of this product
Product Code. 10736182
  • £30.30 / Each
Estimated Shipment: 21-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Precast tweezers
  • PTFE construction
  • With pointed or blunt ends
  • Universal chemical resistance
Specifications
Specifications
Blunt
Precast tweezer
200 mm
Each
PTFE
Tweezers