Learn More
Bohlender™ PTFE Blunt Tip Tweezers
Use these tweezers for a variety of applications. BOLA PTFE Tweezers feature universal chemical resistance.
£24.25 - £30.30
Specifications
Tip Style Blunt
No. per Pack Each
Design Precast tweezer
Material PTFE
Product Type Tweezers
View More Specs
Products 2
Product Code Brand Length (Metric) Price Quantity & Availability  
Product Code Brand Length (Metric) Price Quantity & Availability  
10736182
View Documents
Bohlender™
H912-06
200 mm
£30.30
Each
Estimated Shipment: 20-01-2025
to see stock.
Add to basket
10513832
View Documents
Bohlender™
H912-04
150 mm
£24.25
Each
Estimated Shipment: 20-01-2025
to see stock.
Add to basket
Description
Description
  • Precast tweezers
  • PTFE construction
  • With pointed or blunt ends
  • Universal chemical resistance
Specifications
Specifications
Blunt
Precast tweezer
Tweezers
Each
PTFE