Learn More
Saint Gobain Life Sciences Chemware™ PTFE Boiling Stones
Use these PTFE chips to promote gentle, efficient boiling and help prevent overheating of highly volatile liquids. Chemware™ PTFE Boiling Stones are not affected by acids, hydrocarbons, ketones, esters, alcohols or alkalis.
Brand:  Saint Gobain Life Sciences D1069103
Product Code. 10027750
  • £43.06 / 450g
Estimated Shipment: 19-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Ideal for continuous extraction methods and boiling organic liquids
  • Chemical resistant
  • Minimize superheating
  • Easily cleaned and non-contaminating
  • Shows no trace of face acidity or alkalinity
  • Will not scratch or abrade glassware and leave no residue
  • Usable up to 288°C
  • Stones minimize bumping during reflux and distillation
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Boiling Stone
PTFE
Distillation Columns
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Saint Gobain Life Sciences Chemware™ PTFE Boiling Stones > Chemware™ PTFE Boiling Stones; Material: PTFE