Learn More
Fisherbrand™ PTFE Centrifugal Stirrer Shaft
Use with shaft guides for round vessels with narrow necks. The PTFE centrifugal stir shafts include blades that open during increasing speed, medium to high speeds are required.
£46.05 - £107.64
Specifications
For Use With (Equipment) Universal Stirrer Guide
For Use With (Application) Mixing
Quantity 1 Each
Temperature (Metric) -200°C to +280°C
View More Specs
Products 16
Product Code Brand Diameter (Metric) Shaft Length (Metric) Diameter (Metric) Fan Price Quantity & Availability  
Product Code Brand Diameter (Metric) Shaft Length (Metric) Diameter (Metric) Fan Price Quantity & Availability  
10187701
View Documents
Fisherbrand™
005.04100.12.2
12 mm 1000 mm (EX) 80 mm
£107.00
Each
Estimated Shipment: 21-05-2025
to see stock.
Add to basket
10127801
View Documents
Fisherbrand™
005.0465.12.2
12 mm 650 mm (EX) 80 mm
£94.70
Each
Estimated Shipment: 22-05-2025
to see stock.
Add to basket
10586902
View Documents
Fisherbrand™
005.0465.2
9.5 mm 650 mm (EX) 70 mm
£64.75
Each
Estimated Shipment: 09-06-2025
to see stock.
Add to basket
10268000
View Documents
Fisherbrand™
005.0475.2
9.5 mm 750 mm (EX) 70 mm
£63.02
Each
Estimated Shipment: 22-05-2025
to see stock.
Add to basket
10137801
View Documents
Fisherbrand™
005.0475.12.2
12 mm 750 mm (EX) 80 mm
£84.05
Each
Estimated Shipment: 09-06-2025
to see stock.
Add to basket
10486481
View Documents
Fisherbrand™
005.0465.10.2
10 mm 650 mm (EX) 70 mm
£64.25
Each
Estimated Shipment: 22-05-2025
to see stock.
Add to basket
10035951
View Documents
Fisherbrand™
005.0475.10.2
10 mm 750 mm (EX) 70 mm
£70.75
Each
Estimated Shipment: 06-05-2025
to see stock.
Add to basket
10564362
View Documents
Fisherbrand™
005.430.8.2
8 mm 300 mm 40 mm
£85.55
Each
Estimated Shipment: 20-05-2025
to see stock.
Add to basket
10084650
View Documents
Fisherbrand™
005.440.2
6 mm 400 mm 40 mm
£46.05
Each
Estimated Shipment: 17-06-2025
to see stock.
Add to basket
10467441
View Documents
Fisherbrand™
005.440.8.2
8 mm 400 mm 40 mm
£76.03
Each
Estimated Shipment: 22-05-2025
to see stock.
Add to basket
10626351
View Documents
Fisherbrand™
005.450.2
6 mm 500 mm 50 mm
£67.30
Each
Estimated Shipment: 22-05-2025
to see stock.
Add to basket
10457251
View Documents
Fisherbrand™
005.455.10.2
10 mm 550 mm 70 mm
£107.64
Each
Estimated Shipment: 22-05-2025
to see stock.
Add to basket
10576902
View Documents
Fisherbrand™
005.465.2
9.5 mm 650 mm 70 mm
£56.40
Each
Estimated Shipment: 09-06-2025
to see stock.
Add to basket
10649732
View Documents
Fisherbrand™
005.450.8.2
8 mm 500 mm 50 mm
£62.90
Each
Estimated Shipment: 09-06-2025
to see stock.
Add to basket
10074650
View Documents
Fisherbrand™
005.430.2
6 mm 300 mm 40 mm
£61.89
Each
Estimated Shipment: 30-05-2025
to see stock.
Add to basket
10566902
View Documents
Fisherbrand™
005.455.2
9.5 mm 550 mm 70 mm
£55.80
Each
Estimated Shipment: 30-05-2025
to see stock.
Add to basket
Description
Description
PTFE shaft stirrers have a unique, fully molded construction featuring a rotor molded to a fully PTFE-encapsulated steel reinforced drive shaft.
  • Chemically resistant, inert, and leak-free
  • Easy to clean
  • No cross contamination
  • Shatterproof
  • Non-scratch
  • Use in temperatures up to approximately 280°C
  • Exposed shaft models (EX) are produced with a stainless steel core with an exposed end for more rigid clamping to the chuck drive
  • For round vessels with narrow necks
  • Blades open with increasing speed, medium to high speeds are required
Specifications
Specifications
Universal Stirrer Guide
Mixing
1 Each
-200°C to +280°C
Videos
SDS
Documents
Documents
Certificates
Product Content Correction
Product Title
Fisherbrand™ PTFE Centrifugal Stirrer Shaft