Learn More
Bel-Art™ SP Scienceware™ PTFE Magnetic Stirring Bar Kit
Supplied in a convenient storage box and provide instant availability of PTFE encapsulated Alnico magnets.
Brand: Bel-Art™ F37160-0000
This item is not returnable.
View return policy
Description
- Useful for environmental testing and life science applications in which small sample volumes need to be prepared and evaluated
- Alnico magnet encapsulated in FDA Grade, USP Class VI Teflon™ PTFE low friction coating; durable and inert
- Manufactured in a registered ISO 9001:2015 facility, as verified by SGS certification
- Autoclave safe
Order Info
DimensionsLxW
Specifications
Autoclavable | |
White | |
Vials, Micro test tubes, Centrifuge tubes | |
1/Ea. |
ISO 9001:2015 | |
25.4 x 8mm, 38.1 x 8mm, 50.8 x 8mm (Two Each) | |
Alnico Magnet, PTFE | |
Octagon |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Product Title
Bel-Art™ SP Scienceware™ PTFE Magnetic Stirring Bar Kit > Size: 25.4 x 8mm, 38.1 x 8mm, 50.8 x 8mm
Spot an opportunity for improvement?Share a Content Correction