missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ PTFE Platinum Resistance Probe with 1M Cable

Inert to virtually all chemical reagents. Fisherbrand™ PTFE Thermocouple Type K Probe combine the outstanding properties of PTFE with the accuracy and reliability of platinum resistance and thermocouple thermometry.

Brand:  Fisherbrand™ 500.400.1

 View more versions of this product

Product Code. 11742464

  • £80.90 / Each
Estimated Shipment: 27-01-2025
to see stock.
Explore more special offers
Add to basket

Description

Description

  • Non-contaminating
  • Impervious to chemical diffusion at high pressure
  • Wide operating temperature range, -180 to +300°C
  • Outstanding electrical insulation properties
  • Individually tested
Specifications

Specifications

Probe, PTFE Platinum Resistance
PTFE with Stainless Steel Core
Measuring temperature
50%, 6 to 14 sec.
400 mm
120 g
Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
PRT100 Class A. TCR = 3850 ppm/degree celsius
PTFE Probe Holders and Compression Fittings
-180°C to +280°C
8 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers