missing translation for 'onlineSavingsMsg'
Learn More

Cowie Technology™ PTFE Pointed Tweezers

Cowie Technology™ PTFE Pointed Tweezers are inert to virtually all chemical reagents and completely non-contaminating.

Brand:  Cowie Technology™ 027.101

 View more versions of this product

Product Code. 10292092

  • £18.60 / Each
Estimated Shipment: 17-12-2024
to see stock.
Explore more special offers
Add to basket

Description

Description

  • Chemically inert
  • Non-contaminating
  • Usable to 280°C
  • Manufactured from Virgin PTFE materials with FDA and USP Class VI approvals
Specifications

Specifications

Pointed
PTFE
PTFE Forceps / Tweezers
8 mm
20 mm
2 mm
20 mm
1 Each
100 mm
Smooth
8
10 mm
100 x 20 mm
Rigid
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali