missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cowie Technology™ PTFE Pointed Tweezers
Cowie Technology™ PTFE Pointed Tweezers are inert to virtually all chemical reagents and completely non-contaminating.
£18.60 - £30.45
Specifications
Tip Style | Pointed |
---|---|
No. per Pack | 1 Each |
Material | PTFE |
Product Type | PTFE Forceps / Tweezers |
Finish | Smooth |
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
10292092
|
Cowie Technology™
027.101 |
100 mm |
£18.60
Each |
Estimated Shipment: 08-08-2024
Log in to see stock. |
|||||
10313792
|
Cowie Technology™
027.151 |
150 mm |
£27.80
Each |
Estimated Shipment: 06-08-2024
Log in to see stock. |
|||||
10457742
|
Cowie Technology™
027.201 |
200 mm |
£30.45
Each |
Estimated Shipment: 08-08-2024
Log in to see stock. |
|||||
Description
- Chemically inert
- Non-contaminating
- Usable to 280°C
- Manufactured from Virgin PTFE materials with FDA and USP Class VI approvals
Specifications
Pointed | |
PTFE | |
Smooth |
1 Each | |
PTFE Forceps / Tweezers | |
Rigid |