missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BRAND™ Straight-Bore PTFE Stopcocks
PTFE construction with polypropylene valve lever. Provides outstanding chemical resistance. Includes integrated hose nozzles.
Brand: BRAND™ 88625
Code : A0
Additional Details : Weight : 0.01000kg
Description
- PTFE construction
- Polypropylene valve
- With integrated one-way hose nozzle
Specifications
3 mm | |
6 mm | |
Polypropylene |
PTFE | |
Integrated hose nozzle |