Learn More
BRAND™ Straight-Bore PTFE Stopcocks
PTFE construction with polypropylene valve lever. Provides outstanding chemical resistance. Includes integrated hose nozzles.
Brand:  BRAND™ 88615
35.10 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Product Code. 10569341
  • £37.20 / Each
Estimated Shipment: 20-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • PTFE construction
  • Polypropylene valve
  • With integrated one-way hose nozzle
TRUSTED_SUSTAINABILITY
Specifications
Specifications
2 mm
4 mm
Polypropylene
PTFE
Integrated hose nozzle
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
BRAND™ Straight-Bore PTFE Stopcocks >