missing translation for 'onlineSavingsMsg'
Learn More

Cowie Technology™ PTFE Square-End Tweezers

Cowie Technology™ PTFE Square-End Tweezers are inert to virtually all chemical reagents and completely non-contaminating.

Brand:  Cowie Technology™ 027.150

 View more versions of this product

Product Code. 10510292

  • £26.35 / Each
Estimated Shipment: 11-04-2025
to see stock.
Explore more special offers
Add to basket

Description

Description

  • Chemically inert
  • Non-contaminating
  • Usable to 280°C
  • Manufactured from Virgin PTFE materials with FDA and USP Class VI approvals
Specifications

Specifications

Square
PTFE
PTFE Forceps / Tweezers
10 mm
22.5 mm
5 mm
22.5 mm
1 Each
150 mm
Smooth
10 mm
11 mm
150 x 22.5 mm
Rigid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

One moment while we fetch your results.
Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Cowie Technology™ PTFE Square-End Tweezers > Length: 150 mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.