missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cowie Technology™ PTFE Square-End Tweezers
Cowie Technology™ PTFE Square-End Tweezers are inert to virtually all chemical reagents and completely non-contaminating.
£21.35 - £34.95
Specifications
Tip Style | Square |
---|---|
No. per Pack | 1 Each |
Material | PTFE |
Product Type | PTFE Forceps / Tweezers |
Finish | Smooth |
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
10510292
|
Cowie Technology™
027.150 |
150 mm |
£26.10
Each |
Estimated Shipment: 27-08-2024
Log in to see stock. |
|||||
10281802
|
Cowie Technology™
027.200 |
200 mm |
£34.95
Each |
Estimated Shipment: 29-08-2024
Log in to see stock. |
|||||
10282092
|
Cowie Technology™
027.100 |
100 mm |
£21.35
Each |
Estimated Shipment: 29-08-2024
Log in to see stock. |
|||||
Description
- Chemically inert
- Non-contaminating
- Usable to 280°C
- Manufactured from Virgin PTFE materials with FDA and USP Class VI approvals
Specifications
Square | |
PTFE | |
Smooth | |
Rigid |
1 Each | |
PTFE Forceps / Tweezers | |
5 mm |