missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ PTFE Stopper, Self Releasing

Fisherbrand™ PTFE Stopper, Self Releasing fit standard taper ground glass joints with sealing rings and self releasing ring for ease of removal.

Brand:  Fisherbrand™

Additional Details : Weight : 0.04020kg

 View more versions of this product

Product Code. 16275941

  • £19.65 / Each
Estimated Shipment: 07-10-2024
to see stock.
Add to basket

Description

Description

  • Total Chemical Resistance
  • High Purity
  • Temperature Range -200°C to +280°C
  • Non contaminating
  • Easy to clean
Specifications

Specifications

Fisherbrand™ PTFE Stopper, Self Releasing, Full Length A Type
1
-200°C to +280°C
24/40
65 mm
PTFE
Standard Taper Ground Glass Joints
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers