missing translation for 'onlineSavingsMsg'
Learn More

Cowie Technology™ PTFE T-Type Stopcocks

Cowie Technology™ PTFE T-Type Stopcocks feature a pure PTFE body and plug and combine all the desirable properties of the Ultimate Laboratory Stopcock.

Brand:  Cowie Technology™ 016.104

Additional Details : Weight : 0.10500kg

 View more versions of this product

Product Code. 10078201

  • £25.70 / Each
Estimated Shipment: 13-08-2024
to see stock.
Add to basket

Description

Description

  • Medium vacuum pressure performance
  • Unbreakable
  • Chemically inert
  • Grease free
  • Supplied with borosilicate glass sidearms

Order Info

dimPlugBore

Specifications

Specifications

T-Type
T-Type Stopcock
Autoclavable
1 Each
PTFE
4 mm
PTFE
10 mm
Borosilicate glass sidearms
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers