missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ PTFE Test Tube Rack

Use from cryogenic to 250°C (480°F). Fisherbrand™ PTFE Test Tube Rack is suitable for ovens and high temperature themostats.

Brand:  Fisherbrand™

Code : A0

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 10241852

  • £58.25 / Each
Estimated Shipment: 22-08-2024
to see stock.
Add to basket

Description

Description

  • Inert to virtually all chemical reagents
  • Suitable for repeatedly sterilizing
Specifications

Specifications

Test Tube Rack
Autoclavable
Tubes (13 mm)
Test Tubes
-200°C to +280°C
180 x 60 x 60mm
60 mm
50 mm
60 mm
250 g
Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
White
PTFE
1 Each
180 x 60mm
180 mm
60 mm
5 mm
13 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers