missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ PTFE Test Tube Rack
Use from cryogenic to 250°C (480°F). Fisherbrand™ PTFE Test Tube Rack is suitable for ovens and high temperature themostats.
£58.25 - £66.70
Specifications
Product Type | Test Tube Rack |
---|---|
Certifications/Compliance | Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS) |
Autoclavable | Autoclavable |
Color | White |
Material | PTFE |
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
10241852
|
Fisherbrand™
|
Tubes (13 mm) |
£58.25
Each |
Estimated Shipment: 22-08-2024
Log in to see stock. |
|||||
10467742
|
Fisherbrand™
|
Tubes (8 mm) |
£58.25
Each |
Estimated Shipment: 22-08-2024
Log in to see stock. |
|||||
10241902
|
Fisherbrand™
|
Tubes (19 mm) |
£66.70
Each |
Estimated Shipment: 23-08-2024
Log in to see stock. |
|||||
10659112
|
Fisherbrand™
|
Tubes (30 mm) |
£66.70
Each |
Estimated Shipment: 23-08-2024
Log in to see stock. |
|||||
Description
- Inert to virtually all chemical reagents
- Suitable for repeatedly sterilizing
Specifications
Test Tube Rack | |
Autoclavable | |
PTFE | |
1 Each | |
180 x 60mm | |
60 mm | |
60 mm |
Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS) | |
White | |
Test Tubes | |
-200°C to +280°C | |
180 mm | |
5 mm |