missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ PTFE Test Tube Rack

Use from cryogenic to 250°C (480°F). Fisherbrand™ PTFE Test Tube Rack is suitable for ovens and high temperature themostats.

Brand:  Fisherbrand™ 029.008

 View more versions of this product

Product Code. 10467742

  • £58.85 / Each
Estimated Shipment: 13-02-2025
to see stock.
Explore more special offers
Add to basket

Description

Description

  • Inert to virtually all chemical reagents
  • Suitable for repeatedly sterilizing
Specifications

Specifications

Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
Autoclavable
Tubes (8 mm)
27
1 Each
180 x 60mm
180 mm
45 mm
5 mm
8 mm
Test Tube Rack
White
PTFE
Test Tubes
-200°C to +280°C
180 x 60 x 45mm
60 mm
35 mm
60 mm
250 g
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.