missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ PTFE Thermocouple Type K Probe

Inert to virtually all chemical reagents. Fisherbrand™ PTFE Thermocouple Type K Probe combine the outstanding properties of PTFE with the accuracy and reliability of platinum resistance and thermocouple thermometry.

Brand:  Fisherbrand™

Code : A0

Additional Details : Weight : 0.14000kg

 View more versions of this product

Product Code. 11712474

  • £83.35 / Each
Estimated Shipment: 02-08-2024
to see stock.
Add to basket

Description

Description

  • Non-contaminating
  • Impervious to chemical diffusion at high pressure
  • Wide operating temperature range, -180 to +300°C
  • Outstanding electrical insulation properties
  • Individually tested
Specifications

Specifications

Probe, PTFE Thermocouple Type K
PTFE with Stainless Steel Core
PTFE Probe Holders and Compression Fittings
500 mm
140 g
Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
Measuring temperature
-180°C to +280°C
9 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers