missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ PTFE Thermocouple Type K Probe
Inert to virtually all chemical reagents. Fisherbrand™ PTFE Thermocouple Type K Probe combine the outstanding properties of PTFE with the accuracy and reliability of platinum resistance and thermocouple thermometry.
£70.43 - £105.26
Specifications
Type | Probe, PTFE Thermocouple Type K |
---|---|
Certifications/Compliance | Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS) |
Material | PTFE with Stainless Steel Core |
For Use With (Application) | Measuring temperature |
For Use With (Equipment) | PTFE Probe Holders and Compression Fittings |
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
11792464
|
Fisherbrand™
|
300 mm |
£98.25
Each |
Estimated Shipment: 02-08-2024
Log in to see stock. |
|||||
11772464
|
Fisherbrand™
|
100 mm |
£70.43
Each |
Estimated Shipment: 29-07-2024
Log in to see stock. |
|||||
11782464
|
Fisherbrand™
|
200 mm |
£90.61
Each |
Estimated Shipment: 02-08-2024
Log in to see stock. |
|||||
11702474
|
Fisherbrand™
|
400 mm |
£105.26
Each |
Estimated Shipment: 02-08-2024
Log in to see stock. |
|||||
11712474
|
Fisherbrand™
|
500 mm |
£83.35
Each |
Estimated Shipment: 02-08-2024
Log in to see stock. |
|||||
11722474
|
Fisherbrand™
|
700 mm |
£105.00
Each |
Estimated Shipment: 19-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Non-contaminating
- Impervious to chemical diffusion at high pressure
- Wide operating temperature range, -180 to +300°C
- Outstanding electrical insulation properties
- Individually tested
Specifications
Probe, PTFE Thermocouple Type K | |
PTFE with Stainless Steel Core | |
PTFE Probe Holders and Compression Fittings |
Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS) | |
Measuring temperature | |
-180°C to +280°C |