missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ PTFE Tweezers, Fine Sharp

Fisherbrand™ PTFE Tweezers, Fine Sharp are inert and non-contaminating

Brand:  Fisherbrand™ 027.0201

 View more versions of this product

Product Code. 16206031

  • £18.50 / Each
Estimated Shipment: 07-01-2025
to see stock.
Explore more special offers
Add to basket

Description

Description

  • Total Chemical Resistance
  • High Purity
  • Temperature Range -200°C to +280°C
  • Non contaminating
  • Easy to clean
Specifications

Specifications

PTFE
200 mm
Fisherbrand™ PTFE Tweezers, Fine Sharp
1/Ea.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers