missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ PTFE Tweezers, Fine Square
Fisherbrand™ PTFE Tweezers, Fine Square are inert and non-contaminating
Brand: Fisherbrand™ 027.0150
Description
- Total Chemical Resistance
- High Purity
- Temperature Range -200°C to +280°C
- Non contaminating
- Easy to clean
Specifications
PTFE | |
150 mm | |
Fisherbrand™ PTFE Tweezers, Fine Square | |
1 Ea. |