Learn More
Buerkle™ PTFE Ultrapure Barrel Pump with Discharge Hose
Provides high chemical resistance and safe transfer, even with liquids which attack conventional materials such as PP or PVC.
Brand: Buerkle™ 5606-0601
525.06 GBP valid until 2024-12-31
Use promo code "23267" to get your promotional price.
Code : 03
Additional Details : Weight : 1.37250kg
Description
- Medium-contacting parts manufactured from PTFE
- Suitable for transferring ultra-pure liquids
- Stopcock prevents hazardous vapours from escaping and stops dripping after transfer
- Discharge hose is 1.2m in length
- Immersion tube with a diameter of 32mm
- All valves are made of solid PTFE/PVDF
- Top quality, 3 years’ warranty
Specifications
Immersion depth of 60 cm | |
1 Ea. | |
270 mL/stroke | |
Liquids which attack conventional materials such as Polypropylene or PVC can now be safely transferred (except for elemental Fluorine), transferring ultra-pure liquids |
Discharge Tube | |
PTFE | |
Barrel Pump |