missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GeneBLAzer™ PTGDR CHO-K1 DA Assay Kit
GeneBLAzer™ PTGDR CHO-K1 DA Assay Kit contains the human Prostanoid (PTGDR) receptor.
Brand: GeneBLAzer™ K1373
Code : NEW
Additional Details : Weight : 0.05000kg
Description
- Gene Symbol: PTGDR
- Accession Number: NM_000953.2
- Gene ID: 5729
- Population Status: Single Cell Clone Origin
- GeneBLAzer™ PTGDR CHO-K1 DA (Division Arrested) cells and PTGDR-CRE-bla CHO-K1 cells contain the human Prostanoid (PTGDR) receptor stably integrated into the CellSensor™ CRE-bla CHO-K1 cell line. CellSensor™ CRE-bla CHO-K1 cells (Cat. no.K1129) contain a beta-lactamase reporter gene under control of the Cyclic AMP Response Element (CRE). Division Arrested (DA) cells are available in two configurations: an Assay Kit (which includes cells and sufficient substrate to analyze 1 x 384-well plate), and a tube of cells sufficient to analyze 10 x 384-well plates.
- DA cells are irreversibly division arrested using a low-dose treatment of Mitomycin-C, and have no apparent toxicity or change in cellular signal transduction. Both PTGDR CHO-K1 DA cells and PTGDR-CRE-bla CHO-K1 cells are functionally validated for Z’-factor and EC50concentrations of Prostaglandin D2. In addition, PTGDR-CRE-bla CHO-K1 cells have been tested for assay performance under variable c'nditions, including DMSO concentration, cell number, stimulation time, and substrate loading time (data available upon request). Additional testing data using alternate stimuli are also available.
- The Adenosine A2A Receptor (PTGDR) has been shown to have many roles throughout the body. PTGDR is involved in pain and inflammation regulation, inhibition of platelet aggregation, blood pressure regulation and contributes to ischemic brain damage. Selective antagonists of the PTGDR pathway are being tested as adjuvants to dopaminergic drugs to treat Parkinson's disease and schizophrenia.
Specifications
Store in liquid nitrogen immediately upon receipt or thaw for immediate use. | |
Fluorescent | |
PTGDR CHO-K1 DA cells, LiveBLAzer™-FRET B/G Loading Kit, 70 μg, Solution D, 1 mL, 2 protocols, Certificate of Analysis | |
Division-Arrested Cells Assay Kit |
Gene ID 5729 | |
GPCRs | |
1 Plate |