missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pumice stone, BAKER™, J.T. Baker™
171.00 GBP valid until 2024-12-31
Use promo code "23268" to get your promotional price.
Use promo code "23268" to get your promotional price.
Click to view available options
1 kg
Description
- Acid-soluble substances: max. 6.0%
- Iron (Fe): Passes Test
- Water Soluble Substances: max. 0.20%
Specifications
Packaging | Glass Bottle for Solids |
Quantity | 1 kg |
CAS | 1332-09-8 |
Melting Point | >500°C |
Molecular Formula | C28H37FO7 |
MDL Number | MFCD00148147 |
InChI Key | CIWBQSYVNNPZIQ-UHFFFAOYNA-N |
SMILES | CCC(=O)OCC(=O)C1(OC(=O)CC)C(C)CC2C3CCC4=CC(=O)C=CC4(C)C3(F)C(O)CC12C |
IUPAC Name | 2-[9b-fluoro-10-hydroxy-2,9a,11a-trimethyl-7-oxo-1-(propanoyloxy)-1H,2H,3H,3aH,3bH,4H,5H,7H,9aH,9bH,10H,11H,11aH-cyclopenta[a]phenanthren-1-yl]-2-oxoethyl propanoate |
Molecular Weight (g/mol) | 504.60 |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'