missing translation for 'onlineSavingsMsg'
Learn More
Pumice Stone, Honeywell Fluka™ Product Code.: 15631420

Pumice Stone, Honeywell Fluka™

Product Code. 15631420 Shop All Honeywell Fluka Products
1 kg, Poly Bottle
Click to view available options
Quantity:
500 g
1 kg
Packaging:
Poly Bottle
This item is not returnable. View return policy

Product Code. 15631420

Brand: Honeywell-Fluka 143721KG

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Granulated

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1332-09-8
Molecular Formula C28H37FO7
Molecular Weight (g/mol) 509.626
MDL Number MFCD00148147
InChI Key CIWBQSYVNNPZIQ-YBDIZOPJSA-N
PubChem CID 76973482
IUPAC Name [2-[(8S,9R,10S,11S,13S,16S,17R)-9-fluoro-11-hydroxy-10,13,16-trimethyl-3-oxo-17-propanoyloxy-6,7,8,11,12,14,15,16-octahydrocyclopenta[a]phenanthren-17-yl]-2-oxoethyl] 2,2,3,3,3-pentadeuteriopropanoateShow MoreShow Less
SMILES CCC(=O)OCC(=O)C1(C(CC2C1(CC(C3(C2CCC4=CC(=O)C=CC43C)F)O)C)C)OC(=O)CC

Specifications

Physical Form Granulated
Packaging Poly Bottle
Color Gray
Quantity 1 kg
UN Number NONH for all modes of transport
Chemical Name or Material Pumice Stone
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Pumice Stone, Honeywell Fluka™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.