missing translation for 'onlineSavingsMsg'
Learn More

Labcon™ PurePlus™ 1.2 mL Sample Library Tubes

Sample library tubes come in strips of 8, 12, or as individual tubes.

Brand:  Labcon™ 3911-540-000-9

 View more versions of this product

Product Code. 17537936

  • £56.35 / Pack of 960
Estimated Shipment: 02-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Available in 96-place racks or resealable bags
  • ATP, RNase/DNase, Endotoxin-free (non-pyrogenic)
Specifications

Specifications

Autoclavable
Polypropylene
960/Pk.
12 mL
Clear
Strips of 8, Rack
960 Tubes/Pk., 10 Pk./Cs.
Non-sterile
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers