missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Labcon™ PurePlus™ 1.2 mL Sample Library Tubes
Sample library tubes come in strips of 8, 12, or as individual tubes.
£51.45 - £65.50
Product Code | Brand | Format | Sterility | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Format | Sterility | Price | Quantity & Availability | ||||
17547936
|
Labcon™
3911-545-000-9 |
Strips of 8, Rack | Sterile |
£63.55
Pack of 960 |
In Stock
Log in to see stock. |
||||
17517936
|
Labcon™
3910-540-000-9 |
Individual Tubes, Rack | Non-sterile |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
17527936
|
Labcon™
3910-545-000-9 |
Individual Tubes, Rack | Sterile |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
17537936
|
Labcon™
3911-540-000-9 |
Strips of 8, Rack | Non-sterile |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
17567936
|
Labcon™
3912-545-000-9 |
Strips of 12, Rack | Sterile |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
17557936
|
Labcon™
3912-540-000-9 |
Strips of 12, Rack | Non-sterile |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
17517946
|
Labcon™
3917-800-000-9 |
Strips of 12, Bagged | Non-sterile |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- Available in 96-place racks or resealable bags
- ATP, RNase/DNase, Endotoxin-free (non-pyrogenic)
Specifications
Autoclavable | |
Clear | |
Polypropylene |