Learn More
BRAND™ PVC Burette Clamp
Two burettes up to 50mL can be mounted in seconds. PVC-coated clamps provide a secure grip. Comfortable height adjustment. Graduations remain visible.
Brand: BRAND™ 578000
64.25 GBP valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- Aluminum die casting construction
- With roll mounting
- PVC-coated clamps
- For use with 2 burettes
- Very sturdy design, easy to operate
- Integral support connector
Specifications
Two Burette | |
1 Each | |
Buret Clamp |
PVC-coated Aluminum | |
Roll Mounting |
Product Suggestions
Customers who viewed this item also viewed.
Product Content Correction
Product Title
BRAND™ PVC Burette Clamp > For use with 2 Burettes
Spot an opportunity for improvement?Share a Content Correction