Learn More
Kartell™ Plastilab™ PVC Non-toxic Isoflex Laboratory Hoses
Transparent hoses with markings at every meter for ease of use.
Brand: Kartell™ 0176100
This item is not returnable.
View return policy
Description
- Non-ageing and non-oxiding
- Resistant to both acids and alkalis
- Autoclavable
- Supplied in 20-meter rolls
- Packed in PE bags
Specifications
Autoclavable | |
PVC | |
20 m | |
1 Pack |
2 mm | |
4 mm | |
1 mm | |
Clear |