Learn More
Kartell™ Plastilab™ PVC Non-toxic Isoflex Laboratory Hoses
Transparent hoses with markings at every meter for ease of use.
Brand:  Kartell™ 0176300
 View more versions of this product
Product Code. 11361145
  • £104.00 / 20m
Estimated Shipment: 03-04-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Non-ageing and non-oxiding
  • Resistant to both acids and alkalis
  • Autoclavable
  • Supplied in 20-meter rolls
  • Packed in PE bags
Specifications
Specifications
Autoclavable
PVC
20 m
1 Pack
5 mm
8 mm
1.5 mm
Clear