Learn More
Pyrazine, 98%, Thermo Scientific Chemicals
£26.10 - £84.30
Chemical Identifiers
CAS | 290-37-9 |
---|---|
Molecular Formula | C4H4N2 |
Molecular Weight (g/mol) | 80.09 |
MDL Number | MFCD00006122 |
InChI Key | KYQCOXFCLRTKLS-UHFFFAOYSA-N |
Synonym | 1,4-diazine, p-diazine, paradiazine, piazine, 1,4-diazabenzene, pyrazin, ccris 1331, 1,4-diazin, mixture, mu-pyrazine; |
PubChem CID | 9261 |
ChEBI | CHEBI:30953 |
IUPAC Name | pyrazine |
SMILES | C1=CN=CC=N1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11337178
|
Thermo Scientific Alfa Aesar
016326.09 |
10 g |
£26.10
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11327178
|
Thermo Scientific Alfa Aesar
016326.18 |
50 g |
£84.30
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Pyrazine is used as a flavoring agent. It serves as a component of some herbs in traditional Chinese medicine. It is employed as pharmaceutical intermediates, perfume intermediates and involved in the preparation of other chemicals. In addition to this, it is used in the production of pyrazine 1-oxide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
290-37-9 | |
80.09 | |
KYQCOXFCLRTKLS-UHFFFAOYSA-N | |
9261 | |
pyrazine |
C4H4N2 | |
MFCD00006122 | |
1,4-diazine, p-diazine, paradiazine, piazine, 1,4-diazabenzene, pyrazin, ccris 1331, 1,4-diazin, mixture, mu-pyrazine; | |
CHEBI:30953 | |
C1=CN=CC=N1 |
Specifications
290-37-9 | |
White | |
113°C to 116°C | |
Strong | |
1.5235 | |
UN1325 | |
Hygroscopic | |
1,4-diazine, p-diazine, paradiazine, piazine, 1,4-diazabenzene, pyrazin, ccris 1331, 1,4-diazin, mixture, mu-pyrazine; | |
KYQCOXFCLRTKLS-UHFFFAOYSA-N | |
pyrazine | |
9261 | |
80.09 | |
Crystalline |
50°C to 56°C | |
1.031 g/mL | |
55°C (131°F) | |
C4H4N2 | |
MFCD00006122 | |
103905 | |
14,7957 | |
Soluble in water,alcohol,and ether. | |
C1=CN=CC=N1 | |
80.09 | |
CHEBI:30953 | |
98% | |
Pyrazine |
Safety and Handling
GHS H Statement
H228-H315-H319-H335
Flammable solid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P240-P241-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H228-H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 4.1; Packing Group: III; Proper Shipping Name: FLAMMABLE SOLIDS, ORGANIC, N.O.S.
missing translation for 'einecsNumber' : 206-027-6
missing translation for 'rtecsNumber' : UQ2015000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only