missing translation for 'onlineSavingsMsg'
Learn More

Corning™ PYREX™ Disposable Bulk Glass Serological Pipets, TD, Non-Sterile, Unplugged

Calibrated to deliver (TD) with blow-out. Corning™ Pyrex™ Disposable Bulk Glass Serological Pipets offer long, slender, tapered tips to make pipetting go faster and easier.

Brand:  Corning™ 7079-2N

122.35 GBP valid until 2024-12-31
Use promo code "21936" to get your promotional price.


 View more versions of this product

Product Code. 11912298

  • £142.00 / Pack of 175
Estimated Shipment: 19-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

These disposable glass pipets are calibrated 'To Deliver' with blow-out and offer long, slender, tapered tips to make pipetting go faster and easier. All pipets have negative graduations and fire-polished tips for burr-free, even flow rates. Nonsterile, unplugged, and bulk packed. Reference: ASTM E-714.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Yes
Glass
0.01 mL
0.2 mL
Bulk
2 mL
Single-use
ASTM E714
Yes
11.41 in.
Round
0.06 oz.
Serological Pipet
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers