missing translation for 'onlineSavingsMsg'
Learn More

Pyrex™ Aspirator Bottle with Ground Glass Side Socket And Stopper

Ground neck and supplied with glass stopper

Brand:  Pyrex 1569/16D

244.49 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 11307894

  • £301.00 / Each
Estimated Shipment: 19-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Complies with ISO 4796
  • Ground side socket (size 29/32) to accept stopcock assemblies
  • Ground neck
  • Supplied with glass stopper
TRUSTED_SUSTAINABILITY
Specifications

Specifications

5000 mL
Glass stopper
45/40
Aspirator Bottle
45 mm
Borosilicate Glass
333 mm
181 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
View accessory products
Special Offers

Special Offers