missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyrex™ Aspirator Bottle with Ground Glass Side Socket And Stopper
Ground neck and supplied with glass stopper
Brand: Pyrex 1569/22D
This item is not returnable.
View return policy
Description
- Complies with ISO 4796
- Ground side socket (size 29/32) to accept stopcock assemblies
- Ground neck
- Supplied with glass stopper
Specifications
20,000 mL | |
Glass stopper | |
60/46 | |
Aspirator Bottle |
60 mm | |
Borosilicate Glass | |
508 mm | |
288 mm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction