missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Pyrex™ Borosilicate Glass Erlenmeyer Flask with Quickfit Ground Glass Socket
Grow cultures and store other laboratory reagents in Glass Erlenmeyer flasks. The Pyrex™ borosilicate glass Erlenmeyer flask with QuickFit™ glass socket is sturdy and comes in a variety of sizes.
Brand: Pyrex 1132/14
257.24 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
Grow cultures and store other laboratory reagents in Glass Erlenmeyer flasks. The Pyrex™ borosilicate glass Erlenmeyer flask with QuickFit™ glass socket is sturdy and comes in a variety of sizes.
- Complies to ISO 4797
- Conical flasks with highest quality Quickfit™ ground glass socket
- Borosilicate glass for corrosion free performance and high resistance to thermal shock
- Erlenmeyer flasks with high quality Quickfit™ ground glass socket
- Manufactured from Pyrex™ borosilicate glass for highest resistance to thermal shock and chemical corrosion
- White enamel graduations and marking spot
Specifications
DIN 12387 | |
Borosilicate Glass | |
130 mm | |
24/29 | |
Yes |
250 mL | |
Yes | |
Conical | |
Yes | |
85 mm |