Learn More
Pyrex™ Borosilicate Glass Erlenmeyer Flask with Quickfit Ground Glass Socket
Grow cultures and store other laboratory reagents in Glass Erlenmeyer flasks. The Pyrex™ borosilicate glass Erlenmeyer flask with QuickFit™ glass socket is sturdy and comes in a variety of sizes.
£332.00 - £810.00
Product Code | Brand | Capacity (Metric) | Standard Taper Joint | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Standard Taper Joint | Price | Quantity & Availability | ||||
12622062
|
Pyrex
1132/15 |
250 mL | 29/32 |
£332.00
Pack of 10 |
Estimated Shipment: 17-09-2024
Log in to see stock. |
||||
12463431
|
Pyrex
1132/20 |
500 mL | 29/32 |
£414.00
Pack of 10 |
Estimated Shipment: 05-11-2024
Log in to see stock. |
||||
12453431
|
Pyrex
1132/14 |
250 mL | 24/29 |
£353.98
Pack of 10 |
Estimated Shipment: 22-10-2024
Log in to see stock. |
||||
15597660
|
N/A
1132/02 |
50 mL | 19/26 |
£471.00
Pack of 20 |
Estimated Shipment: 24-09-2024
Log in to see stock. |
||||
15507670
|
N/A
1132/06 |
100 mL | 29/32 |
£452.00
Pack of 20 |
Estimated Shipment: 10-01-2025
Log in to see stock. |
||||
15517670
|
N/A
1132/26 |
1000 mL | 29/32 |
£810.00
Pack of 20 |
Estimated Shipment: 10-01-2025
Log in to see stock. |
||||
Description
Grow cultures and store other laboratory reagents in Glass Erlenmeyer flasks. The Pyrex™ borosilicate glass Erlenmeyer flask with QuickFit™ glass socket is sturdy and comes in a variety of sizes.
- Complies to ISO 4797
- Conical flasks with highest quality Quickfit™ ground glass socket
- Borosilicate glass for corrosion free performance and high resistance to thermal shock
- Erlenmeyer flasks with high quality Quickfit™ ground glass socket
- Manufactured from Pyrex™ borosilicate glass for highest resistance to thermal shock and chemical corrosion
- White enamel graduations and marking spot