Learn More
Pyrex™ Borosilicate Glass Erlenmeyer Flask with Quickfit Ground Glass Socket
Grow cultures and store other laboratory reagents in Glass Erlenmeyer flasks. The Pyrex™ borosilicate glass Erlenmeyer flask with QuickFit™ glass socket is sturdy and comes in a variety of sizes.
Brand: N/A 1132/02
444.48 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : NEW
Additional Details : Weight : 0.37800kg
Description
Grow cultures and store other laboratory reagents in Glass Erlenmeyer flasks. The Pyrex™ borosilicate glass Erlenmeyer flask with QuickFit™ glass socket is sturdy and comes in a variety of sizes.
- Erlenmeyer flasks with high quality Quickfit™ ground glass socket
- White enamel graduations and marking spot
Specifications
50 mL | |
85 mm | |
19/26 | |
50 mm |
Borosilicate Glass | |
10 | |
19/26 |