Learn More
Pyrex™ Borosilicate Glass Erlenmeyer Flask with Quickfit Ground Glass Socket
Grow cultures and store other laboratory reagents in Glass Erlenmeyer flasks. The Pyrex™ borosilicate glass Erlenmeyer flask with QuickFit™ glass socket is sturdy and comes in a variety of sizes.
Brand: Pyrex 1132/20
281.47 GBP valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Code : A0
Additional Details : Weight : 2.53000kg
Description
Grow cultures and store other laboratory reagents in Glass Erlenmeyer flasks. The Pyrex™ borosilicate glass Erlenmeyer flask with QuickFit™ glass socket is sturdy and comes in a variety of sizes.
- Complies to ISO 4797
- Conical flasks with highest quality Quickfit™ ground glass socket
- Borosilicate glass for corrosion free performance and high resistance to thermal shock
- Erlenmeyer flasks with high quality Quickfit™ ground glass socket
- Manufactured from Pyrex™ borosilicate glass for highest resistance to thermal shock and chemical corrosion
- White enamel graduations and marking spot
Specifications
DIN 12387 | |
Borosilicate Glass, Borosilicate Glass | |
167 mm | |
29/32 | |
Yes |
500 mL | |
Yes | |
Conical | |
Yes | |
105 mm |