Learn More
Pyrex™ Quickfit™ Borosilicate Glass Florentine Flask
Remove solid residues with ease, using this robust, pear-shaped flask. Quickfit™ Borosilicate Glass Florentine Flask features a large surface area for rapid evaporation.
Brand: Pyrex™ FD250/4RE
34.60 GBP valid until 2025-01-17
Use promo code "21645" to get your promotional price.
Description
Remove solid residues with ease, using this robust, pear-shaped flask. Quickfit™ Borosilicate Glass Florentine Flask features a large surface area for rapid evaporation.
- Ideal for use with rotary evaporators
- Large surface area for rapid evaporation
- Robust heavy wall construction for added strength
- Shape allows easy removal of solid residues
Specifications
250 mL | |
Borosilicate Glass | |
151 mm | |
29/32 |