Learn More
Pyrex™ Quickfit™ Borosilicate Glass Florentine Flask
Remove solid residues with ease, using this robust, pear-shaped flask. Quickfit™ Borosilicate Glass Florentine Flask features a large surface area for rapid evaporation.
£80.80 - £140.00
Specifications
Material Borosilicate Glass
Standard Taper Joint 29/32
View More Specs
Products 4
Product Code Brand Capacity (Metric) Standard Taper Joint Height (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Standard Taper Joint Height (Metric) Price Quantity & Availability  
12375263
View Documents
Pyrex™
FD1L/4RE
1000 mL 29/32 198 mm
£140.00
Each
In Stock
to see stock.
Edge
Add to basket
11921025
View Documents
Pyrex™
FD100/4RE
100 mL 29/32 107 mm
£80.80
Each
Estimated Shipment: 14-03-2025
to see stock.
Add to basket
12041672
View Documents Promotion Details
Pyrex™
FD250/4RE
250 mL 29/32 151 mm
£115.00
Each
In Stock
to see stock.
Add to basket
12071672
View Documents
Pyrex™
FD500/4RE
500 mL 29/32 160 mm
£135.00
Each
Estimated Shipment: 31-01-2025
to see stock.
Add to basket
Description
Description
Remove solid residues with ease, using this robust, pear-shaped flask. Quickfit™ Borosilicate Glass Florentine Flask features a large surface area for rapid evaporation.
  • Ideal for use with rotary evaporators
  • Large surface area for rapid evaporation
  • Robust heavy wall construction for added strength
  • Shape allows easy removal of solid residues
Specifications
Specifications
Borosilicate Glass
29/32