missing translation for 'onlineSavingsMsg'
Learn More

Pyrex™ Borosilicate Glass Graduated Test Tubes with Stopper

Manufactured with borosilicate glass for excellent chemical and thermal shock resistance. Pyrex™ Borosilicate Glass Graduated Test Tubes with Stopper are ideal for measuring and mixing liquids and solutions.

Brand:  Pyrex 1628/22M

193.51 GBP valid until 2025-01-31
Use promo code "23894" to get your promotional price.


 View more versions of this product

Product Code. 11367814

  • £243.00 / Pack of 10
Estimated Shipment: 24-02-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • With white enamel graduations to indicate capacity
  • Pyrex™ borosilicate glass construction provides excellent resistance to chemical attack and thermal shock
  • Supplied with interchangeable polyethylene stopper
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Polyethylene
200 mm
Interchangeable stopper
Yes
Stopper
50 mL
Borosilicate Glass
10 Pack
26 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers